Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035022.1 | NP_001030194.1 | 6 - 7 | 785 | 110 | |
GenBank mRNA | BC102232.1 | - | 6 - 7 | 785 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |