Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173916.3 | NP_776341.1 | 1 - 2 | 75 | 87 | |
GenBank mRNA | BC102221.1 | - | 1 - 2 | 76 | 87 | |
K00107.1 | - | 1 - 2 | 76 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |