Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174093.1 | NP_776518.1 | 3 - 4 | 172 | 152 | |
GenBank mRNA | EU276067.1 | - | 2 - 3 | 127 | 152 | |
M35589.1 | - | 3 - 4 | 143 | 152 | ||
M37211.1 | - | 3 - 4 | 172 | 152 | ||
X12498.1 | - | 1 | 143 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |