Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174140.3 | NP_776565.1 | 4 - 5 | 661 | 81 | |
GenBank mRNA | AF017058.1 | - | 4 - 5 | 663 | 81 | |
BC112656.1 | - | 5 - 6 | 694 | 81 | ||
U48229.1 | - | 4 - 5 | 651 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |