Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CABP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174255.3 | NP_776680.2 | 3 - 4 | 293 | 72 | |
GenBank mRNA | AF169155.1 | - | 3 - 4 | 293 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Bos taurus Inventoried Probe spans exons |