Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174516.2 | NP_776941.1 | 7 - 8 | 858 | 80 | |
GenBank mRNA | BC147876.1 | - | 7 - 8 | 893 | 80 | |
X00962.1 | - | 858 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |