Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CORO1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174521.2 | NP_776946.1 | 2 - 3 | 292 | 97 | |
GenBank mRNA | BC102876.1 | - | 2 - 3 | 97 | ||
BT025463.1 | - | 2 - 3 | 97 | |||
D44496.1 | - | 2 - 3 | 292 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |