Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABGEF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174591.1 | NP_777016.1 | 4 - 5 | 599 | 128 | |
GenBank mRNA | AJ001119.1 | - | 4 - 5 | 599 | 128 | |
BC109491.1 | - | 4 - 5 | 617 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |