Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP11B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174638.3 | NP_777063.2 | 1 - 2 | 247 | 82 | |
GenBank mRNA | AH003250.2 | - | 1 - 2 | 245 | 82 | |
D00185.1 | - | 247 | 82 |
Gene Symbol: LOC787647 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | D00361.1 | - | 249 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |