Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174830.1 | NP_777255.1 | 3 - 4 | 214 | 78 | |
GenBank mRNA | AF141872.1 | - | 3 - 4 | 214 | 78 | |
BC109922.1 | - | 3 - 4 | 219 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |