Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176659.2 | NP_788832.1 | 1 - 2 | 23 | 58 | |
GenBank mRNA | BC102727.1 | - | 1 - 2 | 55 | 58 | |
X63217.1 | - | 23 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |