Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPK3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012672.1 | NP_001012690.1 | 1 - 2 | 235 | 81 | |
GenBank mRNA | AY233461.1 | - | 1 - 2 | 235 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Bos taurus Probe spans exons |