Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014932.1 | NP_001014932.1 | 3 - 4 | 398 | 83 | |
GenBank mRNA | BC110214.1 | - | 3 - 4 | 409 | 83 | |
BT021538.1 | - | 3 - 4 | 398 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |