Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015598.1 | NP_001015598.1 | 3 - 4 | 1463 | 95 | |
GenBank mRNA | BC146253.1 | - | 3 - 4 | 1467 | 95 | |
BT021600.1 | - | 3 - 4 | 1463 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |