Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCS1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015671.1 | NP_001015671.1 | 6 - 7 | 1204 | 68 | |
GenBank mRNA | BC103135.1 | - | 6 - 7 | 1010 | 68 | |
BT020945.1 | - | 6 - 7 | 1204 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |