Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024487.1 | NP_001019658.1 | 5 - 6 | 1327 | 94 | |
GenBank mRNA | BC119955.1 | - | 5 - 6 | 1279 | 94 | |
BT021763.1 | - | 5 - 6 | 1327 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |