Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRROS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034391.2 | NP_001029563.1 | 1 - 2 | 96 | 125 | |
GenBank mRNA | BC103278.1 | - | 1 - 2 | 98 | 125 | |
BT021150.1 | - | 1 - 2 | 84 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |