Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035294.1 | NP_001030371.1 | 1 - 2 | 290 | 95 | |
GenBank mRNA | BC126586.1 | - | 1 - 2 | 324 | 95 | |
BT021197.1 | - | 1 - 2 | 290 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |