Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHDC8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035370.2 | NP_001030447.2 | 2 - 3 | 564 | 94 | |
GenBank mRNA | BC120240.1 | - | 2 - 3 | 637 | 94 | |
BT020815.1 | - | 2 - 3 | 564 | 94 | ||
BT020938.1 | - | 2 - 3 | 563 | 94 | ||
BT026296.1 | - | 2 - 3 | 554 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |