Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040493.1 | NP_001035583.1 | 6 - 7 | 831 | 150 | |
GenBank mRNA | AB098975.1 | - | 1 - 2 | 177 | 150 | |
BC102824.1 | - | 6 - 7 | 831 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |