Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POFUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043342.2 | NP_001036807.1 | 2 - 3 | 323 | 97 | |
NM_001043343.1 | NP_001036808.1 | 2 - 3 | 325 | 97 | ||
NM_001043344.1 | NP_001036809.1 | 2 - 3 | 325 | 97 | ||
NM_001043345.2 | NP_001036810.1 | 2 - 3 | 323 | 97 | ||
NM_182986.2 | NP_892031.1 | 2 - 3 | 325 | 97 | ||
GenBank mRNA | AJ781503.1 | - | 2 - 3 | 257 | 97 | |
AY344580.2 | - | 2 - 3 | 325 | 97 | ||
AY344581.2 | - | 2 - 3 | 325 | 97 | ||
BC126762.1 | - | 2 - 3 | 298 | 97 | ||
BT021843.1 | - | 2 - 3 | 305 | 97 | ||
DQ138291.1 | - | 2 - 3 | 325 | 97 | ||
DQ138292.1 | - | 2 - 3 | 325 | 97 | ||
DQ138293.1 | - | 2 - 3 | 325 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |