Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GNT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076979.1 | NP_001070447.1 | 1 - 2 | 1471 | 94 | |
GenBank mRNA | BT021862.1 | - | 1 - 2 | 1471 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |