Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCLAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BT021587.1 | - | 5 - 6 | 677 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |