Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174563.3 | NP_776988.2 | 1 - 2 | 165 | 93 | |
GenBank mRNA | AB062594.1 | - | 1 - 2 | 144 | 93 | |
BC112678.1 | - | 1 - 2 | 166 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |