Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174699.2 | NP_777124.1 | 6 - 7 | 615 | 80 | |
GenBank mRNA | AB051629.1 | - | 6 - 7 | 615 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |