Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181001.3 | NP_851344.1 | 1 - 2 | 880 | 72 | |
GenBank mRNA | AF445639.1 | - | 1 - 2 | 882 | 72 | |
AY046527.2 | - | 1 - 2 | 818 | 72 | ||
AY162287.1 | - | 1 - 2 | 873 | 72 | ||
JQ513971.1 | - | 1 - 2 | 818 | 72 | ||
JQ513972.1 | - | 1 - 2 | 818 | 72 | ||
JQ513973.1 | - | 1 - 2 | 818 | 72 | ||
JQ513974.1 | - | 1 - 2 | 818 | 72 | ||
JQ513975.1 | - | 1 - 2 | 818 | 72 | ||
JQ513976.1 | - | 1 - 2 | 818 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Bos taurus Probe spans exons |