Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181811.2 | NP_861527.1 | 1 - 2 | 598 | 57 | |
GenBank mRNA | AB110601.1 | - | 1 - 2 | 519 | 57 | |
AB257561.1 | - | 1 - 2 | 546 | 57 | ||
BC142159.1 | - | 1 - 2 | 598 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |