Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AOC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034361.2 | NP_001029533.1 | 1 - 2 | 57 | 126 | |
GenBank mRNA | BC105152.1 | - | 1 - 2 | 59 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |