Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034402.2 | NP_001029574.1 | 2 - 3 | 237 | 83 | |
GenBank mRNA | BC102320.1 | - | 2 - 3 | 239 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |