Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034662.2 | NP_001029834.1 | 5 - 6 | 485 | 71 | |
GenBank mRNA | BC102206.1 | - | 5 - 6 | 486 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |