Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034783.2 | NP_001029955.1 | 2 - 3 | 147 | 91 | |
GenBank mRNA | BC102748.1 | - | 2 - 3 | 148 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Inventoried Probe spans exons |