Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELOB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034800.1 | NP_001029972.1 | 161 | 72 | ||
GenBank mRNA | BC103196.1 | - | 161 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus |