Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034805.2 | NP_001029977.1 | 4 - 5 | 327 | 152 | |
GenBank mRNA | BC103423.1 | - | 4 - 5 | 328 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |