Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035024.1 | NP_001030196.1 | 3 - 4 | 296 | 74 | |
GenBank mRNA | BC102990.1 | - | 3 - 4 | 296 | 74 | |
BT026310.1 | - | 3 - 4 | 308 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |