Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035314.2 | NP_001030391.1 | 5 - 6 | 448 | 137 | |
GenBank mRNA | BC102893.1 | - | 5 - 6 | 449 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |