Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOSC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035397.2 | NP_001030474.1 | 2 - 3 | 489 | 76 | |
GenBank mRNA | BC102436.1 | - | 2 - 3 | 490 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |