Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037486.2 | NP_001032563.1 | 1 - 2 | 154 | 59 | |
NM_001164221.1 | NP_001157693.1 | 1 - 2 | 154 | 59 | ||
GenBank mRNA | BC108197.1 | - | 1 - 2 | 154 | 59 | |
EV663472.1 | - | 1 - 2 | 136 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |