Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040600.2 | NP_001035690.1 | 1 - 2 | 285 | 90 | |
GenBank mRNA | BC108120.1 | - | 1 - 2 | 286 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |