Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079608.2 | NP_001073076.1 | 1 - 2 | 58 | 111 | |
GenBank mRNA | BC102736.1 | - | 1 - 2 | 59 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |