Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083771.1 | NP_001077240.1 | 2 - 3 | 167 | 57 | |
GenBank mRNA | BC104539.2 | - | 2 - 3 | 167 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |