Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFCAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037585.2 | NP_001032674.1 | 1 - 2 | 148 | 90 | |
GenBank mRNA | BC109799.1 | - | 1 - 2 | 151 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |