Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038048.2 | NP_001033137.1 | 6 - 7 | 1507 | 111 | |
GenBank mRNA | BC109804.1 | - | 6 - 7 | 1508 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |