Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STOML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038068.1 | NP_001033157.1 | 9 - 10 | 1031 | 75 | |
GenBank mRNA | BC109523.1 | - | 9 - 10 | 1031 | 75 | |
BC142027.1 | - | 9 - 10 | 963 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |