Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C29H11orf98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038581.2 | NP_001033670.1 | 1 - 2 | 81 | 65 | |
GenBank mRNA | BC109665.1 | - | 1 - 2 | 82 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |