Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR1C3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038584.1 | NP_001033673.1 | 8 - 9 | 963 | 116 | |
GenBank mRNA | BC109895.1 | - | 8 - 9 | 963 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |