Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046140.2 | NP_001039605.1 | 5 - 6 | 923 | 75 | |
GenBank mRNA | BC105395.1 | - | 5 - 6 | 924 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |