Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAGLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046149.1 | NP_001039614.1 | 4 - 5 | 529 | 66 | |
GenBank mRNA | BC105336.1 | - | 4 - 5 | 529 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |