Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046355.2 | NP_001039820.1 | 6 - 7 | 829 | 71 | |
GenBank mRNA | BC112884.1 | - | 6 - 7 | 830 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |