Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUPT20H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046370.1 | NP_001039835.1 | 20 - 21 | 1800 | 85 | |
GenBank mRNA | BC105571.1 | - | 20 - 21 | 1800 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |