Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB3IL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046532.1 | NP_001039997.1 | 5 - 6 | 803 | 65 | |
GenBank mRNA | BC105507.1 | - | 5 - 6 | 803 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |